missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) bromide, 97%

Catalog No. p-4813352
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC447770250 Supplier Thermo Scientific Chemicals Supplier No. 447770250
Only null left

CAS: 7787-58-8 | BiBr3 | 448.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-58-8
Molecular Formula BiBr3
Molecular Weight (g/mol) 448.69
InChI Key TXKAQZRUJUNDHI-UHFFFAOYSA-K
Synonym bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
PubChem CID 82232
IUPAC Name tribromobismuthane
SMILES Br[Bi](Br)Br

Specifications

Boiling Point 453°C
Melting Point 218°C
Quantity 25 g
Assay Percent Range 96 to 103% (Bi)
Linear Formula BiBr3
Merck Index 15,1262
Solubility Information Solubility in water: reacts. Other solubilities: practically insoluble in alcohol,soluble in solutions of ki,kbr,kcl,dil hcl
Formula Weight 448.69
Percent Purity 97%
Packaging Glass bottle
Chemical Name or Material Bismuth(III) bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 232-121-1

RUO – Research Use Only