missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min

Catalog No. AA4047614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4047614 25 g
AA4047622 100 g
AA4047636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4047614 Supplier Thermo Scientific Chemicals Supplier No. 04047614
Only null left

Ceramic glazes and enamel fluxes | CAS: 1304-85-4

It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1304-85-4
MDL Number MFCD00064844
Synonym Bismuth subnitrate

Specifications

Melting Point 260°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
UN Number UN1477
Merck Index 14,1284
Solubility Information Practically insoluble in water and alcohol; Soluble in dilute HNO3 and HCl
Formula Weight 1461.98
Concentration or Composition (by Analyte or Components) Bi2O3 79% min.
Odor Odorless
Sensitivity Hygroscopic
Density 6.04 g/mL
Chemical Name or Material Bismuth hydroxide nitrate oxide
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 215-136-8
RTECSNumber EB2977000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only