Learn More
Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis)
CAS: 12233-56-6
$314.22 - $1135.10
Description
Bismuth germanium oxide is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12233-56-6 | |
| MFCD00064629 |
Specifications
| White | |
| Powder | |
| MFCD00064629 | |
| 1245.68 | |
| Puratronic | |
| Bismuth germanium oxide |
| 12233-56-6 | |
| 25 g | |
| Insoluble in water. | |
| 99.9995% | |
| Odorless | |
| (metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-457-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only