missing translation for 'onlineSavingsMsg'
Learn More

Bismuth citrate, 94%

Numéro de catalogue. AA40398A1
Modifier l'affichage
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA40398A1 1 kg
AA4039830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA40398A1 Fournisseur Thermo Scientific Chemicals Code fournisseur 040398A1
Il en reste null

CAS: 813-93-4 | C6H5BiO7 | 398.079 g/mol

Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 813-93-4
Molecular Formula C6H5BiO7
Molecular Weight (g/mol) 398.079
MDL Number MFCD00050817
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
Synonym bismuth citrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, bismuth 3+ salt 1:1, bismuth citrate usp:usp, bismuth iii citrate, bismuth citrate usp, bismuth 3+ ion citrate, bismuth, citrato 3-, bismuth iii citrate 250g, bismuth 2-hydroxypropane-1,2,3-tricarboxylate
PubChem CID 13135
IUPAC Name bismuth;2-hydroxypropane-1,2,3-tricarboxylate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Bi+3]

Spécifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Insoluble in water,alcohol,ether; Soluble in ammonia solution and in solution of alkali citrates; Typical tap density 0;50g/cm3;Soluble in ammonia,alkali citrates; Insoluble in water,alcohol and ether.
Formula Weight 398.08
Percent Purity 94%
Odor Odorless
Sensitivity Light sensitive
Density 3.5 g/mL
Chemical Name or Material Bismuth citrate
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 212-390-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only