missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Chloride, 97%, Spectrum™ Chemical

Catalog No. p-7165141
Change view
Click to view available options
Quantity:
100 g
12 kg
25 g
50 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18609089 25 g Amber Glass Bottle
18609087 100 g Amber Glass Bottle
18609090 500 g Amber Glass Bottle
18609088 12 kg Poly Pail
18609091 50 kg Fiber Drum
5 options

Catalog No. 18609089

Supplier: Spectrum Chemical Mfg Cor B111025GM

Only null left

Chemical Identifiers

CAS 7787-60-2

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.