missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Chloride, 97%, Spectrum™ Chemical

Catalog No. 18609089
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609089 25 g Amber Glass Bottle
18609087 100 g Amber Glass Bottle
18609090 500 g Amber Glass Bottle
18609088 12 kg Amber Glass Bottle
18609091 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609089 Supplier Spectrum Chemical Mfg Cor Supplier No. B111025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7787-60-2

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 25 g
Molecular Weight (g/mol) 315.34
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Chloride

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.