missing translation for 'onlineSavingsMsg'
Learn More

Bismuth carbonate oxide

CAS: 5892-10-4 | CBi2O5 | 509.97 g/mol

$64.08 - $575.49

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1248318
View Documents
Thermo Scientific Chemicals
A1248318
50 g
Each for $64.08
Only null left
 
AAA1248330
View Documents
Thermo Scientific Chemicals
A1248330
250 g
Each for $176.53
Only null left
 
AAA124830B
View Documents
Thermo Scientific Chemicals
A124830B
1000 g
Each for $575.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5892-10-4
509.97
MGLUJXPJRXTKJM-UHFFFAOYSA-L
16683095
bis(oxobismuthanyl) carbonate
CBi2O5
MFCD00010877
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
CHEBI:31291
C(=O)(O[Bi]=O)O[Bi]=O

Specifications

5892-10-4
CBi2O5
14,1282
MGLUJXPJRXTKJM-UHFFFAOYSA-L
bis(oxobismuthanyl) carbonate
16683095
509.97
Bismuth carbonate oxide
50 g
MFCD00010877
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
C(=O)(O[Bi]=O)O[Bi]=O
509.97
CHEBI:31291
6.86
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-567-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only