missing translation for 'onlineSavingsMsg'
Learn More

Bismuth carbonate oxide, 98.5% min

Catalog No. AA3321622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3321622 100 g
AA3321636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3321622 Supplier Thermo Scientific Chemicals Supplier No. 03321622
Only null left

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes | CAS: 5892-10-4 | CBi2O5 | 509.97 g/mol

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O

Specifications

Physical Form Powder
Quantity 100 g
Merck Index 14,1282
Solubility Information Practically insoluble in water,alcohol; Soluble in nitric acid,hydrochloric acid,concentrated acetic acid;Soluble in mineral acid and glacial acetic acid; Insoluble in water.
Formula Weight 509.97
Percent Purity ≤98.5%
Density 6.86 g/mL
Chemical Name or Material Bismuth carbonate oxide
EINECSNumber 227-567-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only