missing translation for 'onlineSavingsMsg'
Learn More

Bismuth 2-ethylhexanoate acid

Catalog No. p-7059864
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4550006 5 g
AA4550014 25 g
AA4550022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4550006 Supplier Thermo Scientific Chemicals Supplier No. 04550006

CAS: 67874-71-9 | C24H48BiO6 | 641.622 g/mol

Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67874-71-9
Molecular Formula C24H48BiO6
Molecular Weight (g/mol) 641.622
MDL Number MFCD00043803
InChI Key MWXWNYHYPMRCHN-UHFFFAOYSA-N
Synonym 2-Ethylhexanoic acid bismuth salt
PubChem CID 131857451
IUPAC Name bismuth;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 638.61
Percent Purity 92% (typically)
Physical Form Liquid
Chemical Name or Material Bismuth 2-ethylhexanoate, In 2-ethylhexanoic acid
Hazard Category H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 267-499-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only