missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bismarck Brown R

Catalog No. AAJ6475714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6475714 25 g
AAJ6475722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6475714 Supplier Thermo Scientific Chemicals Supplier No. J6475714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5421-66-9 | C21H26Cl2N8 | 461.395 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5421-66-9
Molecular Formula C21H26Cl2N8
Molecular Weight (g/mol) 461.395
MDL Number MFCD00036386
InChI Key WLKAMFOFXYCYDK-UHFFFAOYSA-N
Synonym C.I. 21010; Basic Brown 4
PubChem CID 79459
IUPAC Name 4-[[3-[(2,4-diamino-5-methylphenyl)diazenyl]-4-methylphenyl]diazenyl]-6-methylbenzene-1,3-diamine;dihydrochloride
SMILES CC1=C(C=C(C=C1)N=NC2=C(C=C(C(=C2)C)N)N)N=NC3=C(C=C(C(=C3)C)N)N.Cl.Cl

Specifications

Melting Point 222°C (decomposition)
Beilstein 1829673
Formula Weight 461.39
Odor Odorless
Color Black to Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bismarck Brown R
EINECSNumber 226-541-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only