missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Bisbenzimide H 33258 Fluorochrome, Trihydrochloride Calbiochem™,

Catalog No. 382061250MG
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
382061250MG 250 mg
382061100MG 100 mg
3820611GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 382061250MG Supplier MilliporeSigma™ Supplier No. 382061250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Membrane-permeable, adenine-thymine-specific fluorescent stain

Chemical Identifiers

CAS 23491-45-4
Molecular Formula C25H37Cl3N6O6
Molecular Weight (g/mol) 623.957
InChI Key URHMSNPWJDDZSH-UHFFFAOYSA-N
Synonym bisbenzimide h 33258 fluorochrome trihydrochloride
PubChem CID 18386398
IUPAC Name 4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride
SMILES CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl

Specifications

Color Yellow
Quantity 250 mg
Solubility Information Water: soluble; Precipitates in phosphate buffered solutions
Formula Weight 533.9
Percent Purity ≥98%
Grade HPLC
Physical Form Solid
Chemical Name or Material Bisbenzimide H 33258 Fluorochrome
Recommended Storage 2° to 8°C (35.6° to 46.4°F)