missing translation for 'onlineSavingsMsg'
Learn More

Bisacrylamide, Molecular Grade, Promega

Catalog No. PRV3143
Encompass
Change view
Click to view available options
Quantity:
125 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3143 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV3143 Supplier Promega Corporation Supplier No. V3143

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cross-linking agent for preparation of polyacrylamide gels.

  • Quality tested and certified free of DNase, RNase and protease activity

Specifications

Chemical Name or Material Bisacrylamide
Physical Form Free-flowing Crystals
Color White
Quantity 125 g
Synonym N, N«-Methylenebisacrylamide F.W. 154.20
Formula Weight 154.2
Grade Molecular Biology