missing translation for 'onlineSavingsMsg'
Learn More

Bis(vinylsulfonyl)methane 96.0+%, TCI America™

$102.03 - $308.03

Chemical Identifiers

CAS 3278-22-6
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.24
MDL Number MFCD03265421
InChI Key IJHIIHORMWQZRQ-UHFFFAOYSA-N
PubChem CID 76764
IUPAC Name (ethenesulfonyl)methanesulfonylethene
SMILES C=CS(=O)(=O)CS(=O)(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B25505G
View Documents
TCI America
B25505G
5 g
Each for $102.03
Only null left
 
B255025G
View Documents
TCI America
B255025G
25 g
Each for $308.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3278-22-6
196.24
IJHIIHORMWQZRQ-UHFFFAOYSA-N
(ethenesulfonyl)methanesulfonylethene
C5H8O4S2
MFCD03265421
76764
C=CS(=O)(=O)CS(=O)(=O)C=C

Specifications

3278-22-6
White
MFCD03265421
IJHIIHORMWQZRQ-UHFFFAOYSA-N
(ethenesulfonyl)methanesulfonylethene
76764
≥96.0% (GC)
Bis(vinylsulfonyl)methane
62°C
C5H8O4S2
5 g
C=CS(=O)(=O)CS(=O)(=O)C=C
196.24
196.24
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3734

TSCA : Yes

SDS
Documents

Documents