missing translation for 'onlineSavingsMsg'
Learn More

Bis[tris(2,4-pentanedionato)titanium(IV)] Hexachlorotitanate(IV), TCI America™

Catalog No. B085525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B085525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B085525G Supplier TCI America Supplier No. B085525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12088-57-2
Molecular Formula C30H42Cl6O12Ti3
MDL Number MFCD00059460
Synonym Bis[tris(acetylacetonato)titanium(IV)] Hexachlorotitanate(IV)

Specifications

Quantity 25 g
Formula Weight 950.96
Physical Form Crystalline Powder
Chemical Name or Material Bis[tris(2,4-pentanedionato)titanium(IV)] Hexachlorotitanate(IV)