missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)palladium(II) Diacetate 98.0+%, TCI America™
$354.17 - $1299.25
Chemical Identifiers
| CAS | 14588-08-0 |
|---|---|
| Molecular Formula | C40H36O4P2Pd |
| Molecular Weight (g/mol) | 749.09 |
| MDL Number | MFCD00010013 |
| InChI Key | YOUIUHDWQIUKAO-UHFFFAOYSA-L |
| Synonym | bis triphenylphosphinepalladium acetate |
| PubChem CID | 73357379 |
| IUPAC Name | (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane) |
| SMILES | CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 14588-08-0 | |
| 749.09 | |
| YOUIUHDWQIUKAO-UHFFFAOYSA-L | |
| 73357379 | |
| CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C40H36O4P2Pd | |
| MFCD00010013 | |
| bis triphenylphosphinepalladium acetate | |
| (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane) |
Specifications
| 14588-08-0 | |
| C40H36O4P2Pd | |
| 1 g | |
| YOUIUHDWQIUKAO-UHFFFAOYSA-L | |
| (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane) | |
| 73357379 | |
| ≥98.0% (T) | |
| Bis(triphenylphosphine)palladium(II) Diacetate |
| Yellow | |
| MFCD00010013 | |
| bis triphenylphosphinepalladium acetate | |
| CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 749.09 | |
| 749.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer