missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) Diacetate 98.0+%, TCI America™

Catalog No. B20421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20421G 1 g
B20425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20421G Supplier TCI America Supplier No. B20421G

Chemical Identifiers

CAS 14588-08-0
Molecular Formula C40H36O4P2Pd
Molecular Weight (g/mol) 749.09
MDL Number MFCD00010013
InChI Key YOUIUHDWQIUKAO-UHFFFAOYSA-L
Synonym bis triphenylphosphinepalladium acetate
PubChem CID 73357379
IUPAC Name (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane)
SMILES CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 749.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(triphenylphosphine)palladium(II) Diacetate
TSCA No
Recommended Storage Freezer