missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl) Malonate 97.0+%, TCI America™

Catalog No. B13025ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13025ML Supplier TCI America Supplier No. B13025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18457-04-0
Molecular Formula C9H20O4Si2
Molecular Weight (g/mol) 248.425
MDL Number MFCD00009631
InChI Key ATCKJLDGNXGLAO-UHFFFAOYSA-N
Synonym bis trimethylsilyl malonate, malonic acid bis trimethylsilyl ester, propanedioic acid, bis trimethylsilyl ester, malonic acid, bis trimethylsilyl ester, malonic acid ditms, malonic acid 2tms, malonic acid, bis-tms, bis trimethylsilyl malonat, malonic acid bis-tms, acmc-1c3vy
PubChem CID 87656
IUPAC Name bis(trimethylsilyl) propanedioate
SMILES C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 mL
UN Number 3272
Formula Weight 248.43
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl) Malonate
TSCA No
Recommended Storage Refrigerator