missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl) malonate, 98%

Catalog No. AC313300100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313300100 10 g Glass bottle
AC313300500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313300100 Supplier Thermo Scientific Chemicals Supplier No. 313300100

CAS: 18457-04-0 | C9H20O4Si2 | 248.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18457-04-0
Molecular Formula C9H20O4Si2
Molecular Weight (g/mol) 248.43
MDL Number MFCD00009631
InChI Key ATCKJLDGNXGLAO-UHFFFAOYSA-N
Synonym bis trimethylsilyl malonate, malonic acid bis trimethylsilyl ester, propanedioic acid, bis trimethylsilyl ester, malonic acid, bis trimethylsilyl ester, malonic acid ditms, malonic acid 2tms, malonic acid, bis-tms, bis trimethylsilyl malonat, malonic acid bis-tms, acmc-1c3vy
PubChem CID 87656
IUPAC Name bis(trimethylsilyl) propanedioate
SMILES C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C

Specifications

Color Colorless to Straw
Density 0.9720g/mL
Boiling Point 63°C to 66°C (1.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4150 to 1.417
Linear Formula (CH3)3SiOC(O)CH2C(O)OSi(CH3)3
Quantity 10 g
Specific Gravity 0.972
Solubility Information Solubility in water: reacts
Formula Weight 248.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl) malonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 242-342-5

RUO – Research Use Only