missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)carbodiimide 98.0+%, TCI America™

Catalog No. p-7119018
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13685G 5 g
B136825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13685G Supplier TCI America Supplier No. B13685G
Only null left

Chemical Identifiers

CAS 1000-70-0
Molecular Formula C7H18N2Si2
Molecular Weight (g/mol) 186.405
MDL Number MFCD00051538
InChI Key KSVMTHKYDGMXFJ-UHFFFAOYSA-N
Synonym bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
PubChem CID 70473
IUPAC Name N,N'-bis(trimethylsilyl)methanediimine
SMILES C[Si](C)(C)N=C=N[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 g
UN Number 1993
Formula Weight 186.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)carbodiimide
RTECSNumber VV1302500
TSCA Yes