missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)acetylene 97.0+%, TCI America™

Catalog No. p-7118846
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B10905G 5 g
B109025G 25 g
2 options

Catalog No. B10905G

Supplier: TCI America B10905G

Only null left

Chemical Identifiers

CAS 14630-40-1
Molecular Formula C8H18Si2
Molecular Weight (g/mol) 170.402
MDL Number MFCD00008276
InChI Key ZDWYFWIBTZJGOR-UHFFFAOYSA-N
Synonym bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
PubChem CID 84564
IUPAC Name trimethyl(2-trimethylsilylethynyl)silane
SMILES C[Si](C)(C)C#C[Si](C)(C)C

Specifications

Color White
Boiling Point 134°C
Quantity 5 g
UN Number 1325
Formula Weight 170.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Bis(trimethylsilyl)acetylene
TSCA Yes
Recommended Storage Refrigerator