missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)acetylene, 99%

Catalog No. p-7081809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1196009 10 g
AAA1196018 50 g
AAA1196030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1196009 Supplier Thermo Scientific Chemicals Supplier No. A1196009

CAS: 14630-40-1 | C8H18Si2 | 170.402 g/mol

BTMSA is used as starting reagent in the synthesis of functionalized 4-R-1,2-bis(trimethylsilyl)benzenes. It is also used in the synthesis of (+)-brasilenyne2 and (β-diketanato)Ag(BTMSA). Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne [P227430]).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BTMSA is used as starting reagent in the synthesis of functionalized 4-R-1,2-bis(trimethylsilyl)benzenes. It is also used in the synthesis of (+)-brasilenyne2 and (β-diketanato)Ag(BTMSA). Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne [P227430]).

Solubility
Not miscible or difficult to mix in water. Soluble in Chloroform and Methanol.

Notes
Store away from oxidizing agents. Do not store together with acids.

Chemical Identifiers

CAS 14630-40-1
Molecular Formula C8H18Si2
Molecular Weight (g/mol) 170.402
MDL Number MFCD00008276
InChI Key ZDWYFWIBTZJGOR-UHFFFAOYSA-N
Synonym bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
PubChem CID 84564
IUPAC Name trimethyl(2-trimethylsilylethynyl)silane
SMILES C[Si](C)(C)C#C[Si](C)(C)C

Specifications

Melting Point 20°C to 24°C
Density 0.77
Boiling Point 134°C to 136°C
Flash Point 2°C (35°F)
Refractive Index 1.427
Linear Formula (CH3)3SiC≡CSi(CH3)3
Quantity 10 g
UN Number UN1993
Beilstein 906870
Solubility Information Not miscible or difficult to mix in water. Soluble in Chloroform and Methanol.
Formula Weight 170.4
Percent Purity 99%
Chemical Name or Material Bis (trimethylsilyl)acetylene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 238-671-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only