missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsiloxy)methylsilane, 97%

CAS: 1873-88-7 | C7H21O2Si3 | 221.498 g/mol

Supplier:  Thermo Scientific Chemicals H60586AK

 View more versions of this product

Catalog No. AAH60586AK


Only null left
Explore available promotions

Description

Description

Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications

Specifications

142°C
1873-88-7
C7H21O2Si3
MFCD00048000
1748455
1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst
SWGZAKPJNWCPRY-UHFFFAOYSA-N
methyl-bis(trimethylsilyloxy)silicon
6327366
97%
0.819
Bis(trimethylsiloxy)methylsilane
250 mL
[(CH3)3SiO]2SiHCH3
UN1993
22°C (72°F)
Miscible with acetone,ethanol,diethyl ether. Insoluble in water.
C[Si](O[Si](C)(C)C)O[Si](C)(C)C
221.498
222.51
1.382

Chemical Identifiers

1873-88-7
221.498
SWGZAKPJNWCPRY-UHFFFAOYSA-N
6327366
C[Si](O[Si](C)(C)C)O[Si](C)(C)C
C7H21O2Si3
MFCD00048000
1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst
methyl-bis(trimethylsilyloxy)silicon
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 217-496-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only