missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsiloxy)methylsilane, 97%

Catalog No. AAH60586AD
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60586AD 50 mL
AAH60586AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60586AD Supplier Thermo Scientific Chemicals Supplier No. H60586AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1873-88-7 | C7H21O2Si3 | 221.498 g/mol

Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 1873-88-7
Molecular Formula C7H21O2Si3
Molecular Weight (g/mol) 221.498
MDL Number MFCD00048000
InChI Key SWGZAKPJNWCPRY-UHFFFAOYSA-N
Synonym 1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst
PubChem CID 6327366
IUPAC Name methyl-bis(trimethylsilyloxy)silicon
SMILES C[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 142°C
Quantity 50 mL
Flash Point 22°C (72°F)
Refractive Index 1.382
Density 0.819
Linear Formula [(CH3)3SiO]2SiHCH3
UN Number UN1993
Beilstein 1748455
Solubility Information Miscible with acetone,ethanol,diethyl ether. Insoluble in water.
Formula Weight 222.51
Percent Purity 97%
Chemical Name or Material Bis(trimethylsiloxy)methylsilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 217-496-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only