missing translation for 'onlineSavingsMsg'
Learn More

Bis(trifluoro-2,4-pentanedionato)cobalt(II) Hydrate 90.0+%, TCI America™

Catalog No. T07465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07465G Supplier TCI America Supplier No. T07465G
Only null left

Chemical Identifiers

CAS 16092-38-9
Molecular Formula C10H10CoF6O4
Molecular Weight (g/mol) 367.11
MDL Number MFCD00044994
InChI Key GCHODJIKBYKCNY-UHFFFAOYSA-N
Synonym Cobalt(II) Trifluoroacetylacetonate, Trifluoroacetylacetono Cobalt(II) Salt
PubChem CID 102601710
IUPAC Name bis(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) cobalt
SMILES [Co].CC(=O)C=C(O)C(F)(F)F.CC(=O)C=C(O)C(F)(F)F

Specifications

Color Red-Yellow
Quantity 5 g
Formula Weight 365.09
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trifluoro-2,4-pentanedionato)cobalt(II) Hydrate