missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(0), 98%

Catalog No. AC363511000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363511000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC363511000 Supplier Thermo Scientific Chemicals Supplier No. 363511000

CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33309-88-5
Molecular Formula C36H66P2Pd
Molecular Weight (g/mol) 667.29
MDL Number MFCD01073796
InChI Key JGBZTJWQMWZVNX-UHFFFAOYSA-N
Synonym bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o
PubChem CID 2734559
IUPAC Name palladium;tricyclohexylphosphane
SMILES [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble in water
Formula Weight 667.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(tricyclohexylphosphine)palladium(0)
missing translation for 'healthHazard1' AIR SENSITIVE

RUO – Research Use Only