missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

Catalog No. AAL1801303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1801303 1 g
AAL1801306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1801303 Supplier Thermo Scientific Chemicals Supplier No. L1801303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 164298-25-3 | C9H16F7N2P | 316.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 164298-25-3
Molecular Formula C9H16F7N2P
Molecular Weight (g/mol) 316.204
MDL Number MFCD02683430
InChI Key MNJUGQKOHJQOCK-UHFFFAOYSA-N
Synonym btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
PubChem CID 10935980
IUPAC Name 1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼155°C
Quantity 1 g
UN Number UN1759
Sensitivity Moisture sensitive
Formula Weight 316.2
Chemical Name or Material Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only