missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetrabutylammonium) Bis(maleonitriledithiolato)nickel(II) Complex 97.0+%, TCI America™

Catalog No. B13711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13711G Supplier TCI America Supplier No. B13711G

Chemical Identifiers

CAS 18958-57-1
Molecular Formula C40H72N6NiS4
Molecular Weight (g/mol) 823.991
MDL Number MFCD00059041
InChI Key KFDRXQXGGQXFNP-DERJAXIWSA-J
PubChem CID 11445800
IUPAC Name (Z)-1,2-dicyanoethene-1,2-dithiolate;nickel(2+);tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.C(#N)C(=C(C#N)[S-])[S-].C(#N)C(=C(C#N)[S-])[S-].[Ni+2]

Specifications

Melting Point 142°C
Quantity 1 g
UN Number 3439
Formula Weight 823.99
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tetrabutylammonium) Bis(maleonitriledithiolato)nickel(II) Complex