missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc Complex 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 174°C |
| Color | Purple-Red |
| Quantity | 5 g |
| Formula Weight | 942.99 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc Complex [Organic Electronic Ma |
Safety and Handling
| TSCA | No |