missing translation for 'onlineSavingsMsg'
Learn More

(+)-Bis[(R)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%

Catalog No. AAH2742414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2742414 25 g
AAH2742403 1 g
AAH2742406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2742414 Supplier Thermo Scientific Chemicals Supplier No. H2742414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23294-41-9 | C16H19N | 225.34 g/mol

(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 23294-41-9
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.34
MDL Number MFCD00243088
InChI Key NXLACVVNHYIYJN-UHFFFAOYNA-N
Synonym +-bis r-1-phenylethyl amine, r-bis r-1-phenylethyl amine, bis r-alpha-methylbenzyl amine, +-bis r-1-phenylethy amine hydrochloride, 1r-1-phenyl-n-1r-1-phenylethyl ethanamine, bis 1r-1-phenylethyl amine, r,r-bis-1-phenylethyl amin, +-bis-r-1-phenylethyl amine hydrochloride
PubChem CID 5702599
IUPAC Name (1R)-1-phenyl-N-[(1R)-1-phenylethyl]ethanamine
SMILES CC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 98°C (0.3 mmHg)
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 225.34
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (+)-Bis[(R)-1-phenylethyl]amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only