missing translation for 'onlineSavingsMsg'
Learn More

Bis(pinacolato)diboron 99.0+%, TCI America™

Catalog No. p-7119385
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19641G 1 g
B19645G 5 g
B196425G 25 g
B1964100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B19641G Supplier TCI America Supplier No. B19641G
Only null left

Chemical Identifiers

CAS 73183-34-3
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
MDL Number MFCD00799570
InChI Key IPWKHHSGDUIRAH-UHFFFAOYSA-N
Synonym bis pinacolato diboron, bis pinacolato diborane, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi 1,3,2-dioxaborolane, pinacol diborane, diboron pinacol ester, b2pin2, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3,2-dioxaborolane, bispinacolate diboron, unii-i906w26p4u
PubChem CID 2733548
IUPAC Name 4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
Formula Weight 253.94
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(pinacolato)diboron