Learn More
Bis(pentamethylcyclopentadienyl)zirconium dichloride, 99%, Thermo Scientific Chemicals
$446.62 - $446.62
Chemical Identifiers
| CAS | 54039-38-2 |
|---|---|
| Molecular Formula | C20H30Cl2Zr-2 |
| Molecular Weight (g/mol) | 432.584 |
| MDL Number | MFCD00058849 |
| InChI Key | OCFSLQKTBFSWPL-UHFFFAOYSA-L |
| Synonym | decamethylzirconocene dichloride |
| PubChem CID | 57369741 |
| IUPAC Name | dichlorozirconium;1,2,3,4,5-pentamethylcyclopenta-1,3-diene |
| SMILES | C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.Cl[Zr]Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54039-38-2 | |
| 432.584 | |
| OCFSLQKTBFSWPL-UHFFFAOYSA-L | |
| 57369741 | |
| C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.Cl[Zr]Cl |
| C20H30Cl2Zr-2 | |
| MFCD00058849 | |
| decamethylzirconocene dichloride | |
| dichlorozirconium;1,2,3,4,5-pentamethylcyclopenta-1,3-diene |
Specifications
| 54039-38-2 | |
| Ampule | |
| MFCD00058849 | |
| UN3096 | |
| decamethylzirconocene dichloride | |
| C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.Cl[Zr]Cl | |
| 432.584 | |
| 432.56 | |
| Crystalline |
| Yellow | |
| C20H30Cl2Zr-2 | |
| 2 g | |
| Air and moisture sensitive | |
| OCFSLQKTBFSWPL-UHFFFAOYSA-L | |
| dichlorozirconium;1,2,3,4,5-pentamethylcyclopenta-1,3-diene | |
| 57369741 | |
| 99% | |
| Bis(pentamethylcyclopentadienyl)zirconium dichloride |
Safety and Handling
GHS H Statement
H261-H301-H311-H314-H318-H332
In contact with water releases flammable gas.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only