missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals

$208.30 - $208.30

Chemical Identifiers

CAS 67506-86-9
MDL Number MFCD00058844
Synonym Decamethylmanganocene
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3860503
View Documents
Thermo Scientific Chemicals
03860503
1 g
Each for $208.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67506-86-9
MFCD00058844
Decamethylmanganocene

Specifications

67506-86-9
Ampule
1 g
Decamethylmanganocene
98%
Bis(pentamethylcyclopentadienyl)manganese
230°C (decomposition)
MFCD00058844
Air and moisture sensitive
325.38
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only