missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals

Catalog No. AA3860503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3860503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3860503 Supplier Thermo Scientific Chemicals Supplier No. 03860503
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67506-86-9
MDL Number MFCD00058844
Synonym Decamethylmanganocene

Specifications

Melting Point 230°C (decomposition)
Packaging Ampule
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 325.38
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)manganese
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only