missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-tolyl)phosphine oxide, 98%

Catalog No. p-7041258
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6150403 1 g
AAH6150406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6150403 Supplier Thermo Scientific Chemicals Supplier No. H6150403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2409-61-2 | C14H14OP+ | 229.239 g/mol

Bis(p-tolyl)phosphine oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(p-tolyl)phosphine oxide is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2409-61-2
Molecular Formula C14H14OP+
Molecular Weight (g/mol) 229.239
MDL Number MFCD01445489
InChI Key ZHIPXAFNKGZMSC-UHFFFAOYSA-N
Synonym bis p-tolyl phosphine oxide, di-p-tolylphosphine oxide, bis 4-methylphenyl phosphine oxide, di p-tolyl phosphine oxide, phosphine oxide, bis 4-methylphenyl, 1-methyl-4-4-methylphenylphosphoroso benzene, bis-p-tolylphosphine oxide, oxo bis-p-tolyl phosphonium, bis 4-tolyl phosphine oxide, ksc916e3n
PubChem CID 13357841
IUPAC Name bis(4-methylphenyl)-oxophosphanium
SMILES CC1=CC=C(C=C1)[P+](=O)C2=CC=C(C=C2)C

Specifications

Quantity 1 g
Beilstein 2804632
Solubility Information Slightly soluble in water.
Formula Weight 230.29
Percent Purity 98%
Chemical Name or Material Bis(p-tolyl)phosphine oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only