missing translation for 'onlineSavingsMsg'
Learn More

Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%

Catalog No. p-3212441
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC382551000 100 mg Glass bottle
AC382555000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC382551000 Supplier Thermo Scientific Chemicals Supplier No. 382551000
Only null left

CAS: 36620-11-8 | C14H16BF4Rh | 373.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36620-11-8
Molecular Formula C14H16BF4Rh
Molecular Weight (g/mol) 373.99
MDL Number MFCD00671775
InChI Key HAYDJWBQWOEERB-UHFFFAOYSA-N
Synonym bis norbornadiene rhodium i tetrafluoroborate, rh nbd 2bf4, bis eta-2,5-norbornadiene rhodium i tetrafluoroborate, bis norbornadiene tetrafluoroborato rhodium, bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate, rh nbd 2 bf4, bis norbomadiene rhodium i tetrafluoroborate, bis norbornadiene rhodium tetrafluoroborate
PubChem CID 10915722
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
SMILES [Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Melting Point 209.0°C to 211.0°C
Color Red
Elemental Analysis 92% min.
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: decomposes
Formula Weight 373.99
Percent Purity 94%
Physical Form Powder
Chemical Name or Material Bis(norbornadiene)rhodium(I) tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only