missing translation for 'onlineSavingsMsg'
Learn More

Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate 97.0+%, TCI America™

Catalog No. b250925g
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B250925G 25 g
B2509250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B250925G Supplier TCI America Supplier No. B250925G

Chemical Identifiers

CAS 75381-80-5
Molecular Formula C8H19Br3N2O2
Synonym Bis(N,N-dimethylacetamide) Hydrogen Tribromide

Specifications

Melting Point 92°C
Quantity 25 g
Formula Weight 414.96
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate