missing translation for 'onlineSavingsMsg'
Learn More

Bis(methyldiphenylphosphine)palladium(II) Dichloride 96.0+%, TCI America™

Catalog No. p-7119517
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21611G Supplier TCI America Supplier No. B21611G
Only null left

Chemical Identifiers

CAS 52611-08-2
Molecular Formula C26H26Cl2P2Pd
Molecular Weight (g/mol) 577.762
MDL Number MFCD00216594
InChI Key GTSFBKGHQLDFHU-UHFFFAOYSA-L
Synonym Dichlorobis(methyldiphenylphosphine)palladium(II)
PubChem CID 6102084
IUPAC Name dichloropalladium;methyl(diphenyl)phosphane
SMILES CP(C1=CC=CC=C1)C2=CC=CC=C2.CP(C1=CC=CC=C1)C2=CC=CC=C2.Cl[Pd]Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 577.76
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(methyldiphenylphosphine)palladium(II) Dichloride