missing translation for 'onlineSavingsMsg'
Learn More

Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) 80+%, TCI America™

Catalog No. B4373200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4373200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4373200MG Supplier TCI America Supplier No. B4373200MG
Only null left

Chemical Identifiers

CAS 502693-09-6
Molecular Formula C42H52N6O4RuS2
MDL Number MFCD12546029
Synonym Z907 Dye

Specifications

Melting Point 307°C
Color Black
Quantity 200 mg
Formula Weight 870.11
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(isothiocyanato)(2,2′-bipyridyl-4,4′-dicarboxylato)(4,4′-dinonyl-2,2′-bip