missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexylene Glycolato)diboron 98.0+%, TCI America™

Catalog No. B29061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29061G 1 g
B29065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29061G Supplier TCI America Supplier No. B29061G

Chemical Identifiers

CAS 230299-21-5
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
MDL Number MFCD03490499
InChI Key UEBSWKNVDRJVHN-UHFFFAOYSA-N
Synonym bis hexylene glycolato diboron, bis hexyleneglycolato diboron, 4,4,6-trimethyl-2-4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl, 4,4,4',4',6,6'-hexamethyl-2,2'-bi-1,3,2-dioxaborinane, pubchem7774
PubChem CID 2734616
IUPAC Name 4,4,6-trimethyl-2-(4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl)-1,3,2-dioxaborinane
SMILES B1(OC(CC(O1)(C)C)C)B2OC(CC(O2)(C)C)C

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 253.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(hexylene Glycolato)diboron