missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexafluoroacetylacetonato)manganese(II) Hydrate 95.0+%, TCI America™

$115.57 - $350.44

Chemical Identifiers

CAS 19648-86-3
Molecular Formula C10H4F12MnO4
Molecular Weight (g/mol) 471.06
MDL Number MFCD00042513
InChI Key DMABZFSNYKYMMQ-UHFFFAOYSA-N
Synonym Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate
PubChem CID 91658961
IUPAC Name bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese
SMILES [Mn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H05571G
View Documents
TCI America
H05571G
1 g N/A
Only null left
 
H05575G
View Documents
TCI America
H05575G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

19648-86-3
471.06
DMABZFSNYKYMMQ-UHFFFAOYSA-N
91658961
[Mn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
C10H4F12MnO4
MFCD00042513
Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate
bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese

Specifications

19648-86-3
MFCD00042513
Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate
[Mn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
471.06
469.04
Crystalline Powder
C10H4F12MnO4
1 g
DMABZFSNYKYMMQ-UHFFFAOYSA-N
bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese
91658961
≥95.0% (T)
Bis(hexafluoroacetylacetonato)manganese(II) Hydrate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'