missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexafluoroacetylacetonato)manganese(II) Hydrate 95.0+%, TCI America™

Catalog No. H05575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05575G 5 g
H05571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05575G Supplier TCI America Supplier No. H05575G
Only null left

Chemical Identifiers

CAS 19648-86-3
Molecular Formula C10H4F12MnO4
Molecular Weight (g/mol) 471.06
MDL Number MFCD00042513
InChI Key DMABZFSNYKYMMQ-UHFFFAOYSA-N
Synonym Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate
PubChem CID 91658961
IUPAC Name bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese
SMILES [Mn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 469.04
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(hexafluoroacetylacetonato)manganese(II) Hydrate