missing translation for 'onlineSavingsMsg'
Learn More

Bis(heptafluoroisopropyl)ketone, 97%

Catalog No. AAL1670806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1670806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1670806 Supplier Thermo Scientific Chemicals Supplier No. L1670806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 813-44-5 | C7F14O | 366.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 813-44-5
Molecular Formula C7F14O
Molecular Weight (g/mol) 366.054
MDL Number MFCD00042087
InChI Key GRVMOMUDALILLH-UHFFFAOYSA-N
Synonym bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
PubChem CID 69941
IUPAC Name 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
SMILES C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F

Specifications

Density 1.661
Boiling Point 72°C to 73°C
Quantity 5 g
UN Number UN2810
Beilstein 1916532
Formula Weight 366.06
Percent Purity 97%
Chemical Name or Material Bis(heptafluoroisopropyl)ketone
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 212-384-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only