missing translation for 'onlineSavingsMsg'
Learn More

Bis(ethylenedithio)tetrathiafulvalene

Catalog No. p-7050827
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39557MD 250 mg
AA3955703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39557MD Supplier Thermo Scientific Chemicals Supplier No. 039557MD
Only null left

CAS: 66946-48-3 | C10H8S8 | 384.65 g/mol

It is used as a component for organic superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 66946-48-3
Molecular Formula C10H8S8
Molecular Weight (g/mol) 384.65
MDL Number MFCD00059710
InChI Key LZJCVNLYDXCIBG-UHFFFAOYSA-N
Synonym bis ethylenedithio tetrathiafulvalene, bedt-ttf, 2-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiine, bis ethylenedithio tetrathiofulvalene, bis ethylenedithiolo tetrathiafulvalene, bis-ethylenedithio tetrathiafulvalene, bis ethylenedithiolo tetrathiafulvalene 250mg, bi 5,6-dihydro-1,3-dithiolo 4,5-b 1,4-dithiine-2-ylidene, 2-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiine
PubChem CID 633432
IUPAC Name 2-(5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene)-5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine
SMILES C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1

Specifications

Melting Point 238°C (decomposition)
Quantity 250 mg
Solubility Information Soluble in organic solvents.
Formula Weight 384.68
Physical Form Powder
Chemical Name or Material Bis(ethylenedithiolo)tetrathiafulvalene
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only