missing translation for 'onlineSavingsMsg'
Learn More

Bis(ethylenedithio)tetrathiafulvalene [Organic Electronic Material], TCI America™

Catalog No. B1200100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1200100MG 100 mg
B12001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1200100MG Supplier TCI America Supplier No. B1200100MG

Chemical Identifiers

CAS 66946-48-3
Molecular Formula C10H8S8
Molecular Weight (g/mol) 384.65
MDL Number MFCD00059710
InChI Key LZJCVNLYDXCIBG-UHFFFAOYSA-N
Synonym bis ethylenedithio tetrathiafulvalene, bedt-ttf, 2-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiine, bis ethylenedithio tetrathiofulvalene, bis ethylenedithiolo tetrathiafulvalene, bis-ethylenedithio tetrathiafulvalene, bis ethylenedithiolo tetrathiafulvalene 250mg, bi 5,6-dihydro-1,3-dithiolo 4,5-b 1,4-dithiine-2-ylidene, 2-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiine
PubChem CID 633432
IUPAC Name 2-{2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene}-2H,5H,6H-[1,3]dithiolo[4,5-b][1,4]dithiine
SMILES C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1

Specifications

Quantity 100 mg
Formula Weight 384.65
Physical Form Crystalline Powder
Chemical Name or Material Bis(ethylenedithio)tetrathiafulvalene [Organic Electronic Material]