missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(dithiobenzil)nickel(II) 95.0+%, TCI America™
Specifications
Specifications
| Color | Black-Green |
| Quantity | 1 g |
| Formula Weight | 543.40 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(dithiobenzil)nickel(II) |
Safety and Handling
| TSCA | Yes |