missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(dimethylamino)phosphoryl Chloride 93.0+%, TCI America™
$137.94 - $352.30
Chemical Identifiers
| CAS | 1605-65-8 |
|---|---|
| Molecular Formula | C4H12ClN2OP |
| Molecular Weight (g/mol) | 170.58 |
| MDL Number | MFCD00008302 |
| InChI Key | WYLQARGYFXBZMD-UHFFFAOYSA-N |
| Synonym | bis dimethylamino phosphoryl chloride, phosphorodiamidic chloride, tetramethyl, bis dimethylamino phosphinic chloride, n,n,n',n'-tetramethylphosphorodiamidic chloride, bis dimethylamino phosphorylchloride, bis n,n-dimethylamino phosphinic chloride, ch3 2n 2pocl, bis dimethylamino phosphorochloridate, tetramethylphosphorodiamidic chloride, phosphorodiamidicchloride, n,n,n',n'-tetramethyl |
| PubChem CID | 74150 |
| IUPAC Name | bis(dimethylamino)phosphinoyl chloride |
| SMILES | CN(C)P(Cl)(=O)N(C)C |
Chemical Identifiers
| 1605-65-8 | |
| 170.58 | |
| WYLQARGYFXBZMD-UHFFFAOYSA-N | |
| 74150 | |
| CN(C)P(Cl)(=O)N(C)C |
| C4H12ClN2OP | |
| MFCD00008302 | |
| bis dimethylamino phosphoryl chloride, phosphorodiamidic chloride, tetramethyl, bis dimethylamino phosphinic chloride, n,n,n',n'-tetramethylphosphorodiamidic chloride, bis dimethylamino phosphorylchloride, bis n,n-dimethylamino phosphinic chloride, ch3 2n 2pocl, bis dimethylamino phosphorochloridate, tetramethylphosphorodiamidic chloride, phosphorodiamidicchloride, n,n,n',n'-tetramethyl | |
| bis(dimethylamino)phosphinoyl chloride |
Specifications
| 1605-65-8 | |
| 110°C | |
| MFCD00008302 | |
| 3265 | |
| WYLQARGYFXBZMD-UHFFFAOYSA-N | |
| bis(dimethylamino)phosphinoyl chloride | |
| 74150 | |
| ≥93.0% (GC) | |
| Bis(dimethylamino)phosphoryl Chloride |
| Yellow | |
| C4H12ClN2OP | |
| 5 g | |
| bis dimethylamino phosphoryl chloride, phosphorodiamidic chloride, tetramethyl, bis dimethylamino phosphinic chloride, n,n,n',n'-tetramethylphosphorodiamidic chloride, bis dimethylamino phosphorylchloride, bis n,n-dimethylamino phosphinic chloride, ch3 2n 2pocl, bis dimethylamino phosphorochloridate, tetramethylphosphorodiamidic chloride, phosphorodiamidicchloride, n,n,n',n'-tetramethyl | |
| CN(C)P(Cl)(=O)N(C)C | |
| 170.58 | |
| 170.58 | |
| Liquid |
Safety and Handling
TSCA : No