missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)dimethylsilane, 97%, Thermo Scientific Chemicals

Catalog No. p-7025571
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2111922 Supplier Thermo Scientific Chemicals Supplier No. B2111922
Only null left

Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.

Chemical Identifiers

CAS 3768-58-9
Molecular Formula C6H18N2Si
Molecular Weight (g/mol) 146.309
MDL Number MFCD00008296
InChI Key QULMGWCCKILBTO-UHFFFAOYSA-N
Synonym bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine
PubChem CID 77384
IUPAC Name N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine
SMILES CN(C)[Si](C)(C)N(C)C

Specifications

Boiling Point 128°C to 129°C
Melting Point -98°C
Quantity 100 g
UN Number UN2924
Beilstein 1736095
Flash Point −7°C (19°F)
Solubility Information Hydrolyzes in water.
Formula Weight 146.31
Percent Purity 97%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.807
Chemical Name or Material Bis(dimethylamino)dimethylsilane
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 223-200-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only