missing translation for 'onlineSavingsMsg'
Learn More

Bis(diisopropylamino)chlorophosphine

Catalog No. AC321560250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321560250 25 g Glass bottle
AC321560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC321560250 Supplier Thermo Scientific Chemicals Supplier No. 321560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56183-63-2 | C12H28ClN2P | 266.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56183-63-2
Molecular Formula C12H28ClN2P
Molecular Weight (g/mol) 266.79
MDL Number MFCD00061482
InChI Key FEHUTHGOLLQBNW-UHFFFAOYSA-N
Synonym bis diisopropylamino chlorophosphine, phosphorodiamidous chloride, tetrakis 1-methylethyl, chloro diisopropylamino phosphanyl diisopropylamine, pubchem6478, acmc-1av77, bisdiisopropylaminochlorophosphine, bis diisopropyamino chlorophosphine, bis di-isopropylamino chlorophosphine, bis n,n-diisopropylamino chlorophosphine, chlorobis n,n-diisopropylamino phosphine
PubChem CID 2733425
IUPAC Name N-[chloro-[di(propan-2-yl)amino]phosphanyl]-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C

Specifications

Melting Point 100.0°C to 110.0°C
Packaging Glass bottle
Linear Formula [((CH3)2CH)2N]2PCl
Quantity 25 g
Solubility Information Solubility in water: reacts
Formula Weight 266.8
Chemical Name or Material Bis(diisopropylamino)chlorophosphine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only