missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(diethylamino)dimethylsilane 98.0+%, TCI America™
$71.23 - $209.84
Chemical Identifiers
| CAS | 4669-59-4 |
|---|---|
| Molecular Formula | C10H26N2Si |
| Molecular Weight (g/mol) | 202.417 |
| MDL Number | MFCD00039879 |
| InChI Key | XIFOKLGEKUNZTI-UHFFFAOYSA-N |
| Synonym | N,N,N′,N′-Tetraethyl-1,1-dimethylsilanediamine |
| PubChem CID | 78399 |
| IUPAC Name | N-[diethylamino(dimethyl)silyl]-N-ethylethanamine |
| SMILES | CCN(CC)[Si](C)(C)N(CC)CC |
Chemical Identifiers
| 4669-59-4 | |
| 202.417 | |
| XIFOKLGEKUNZTI-UHFFFAOYSA-N | |
| 78399 | |
| CCN(CC)[Si](C)(C)N(CC)CC |
| C10H26N2Si | |
| MFCD00039879 | |
| N,N,N′,N′-Tetraethyl-1,1-dimethylsilanediamine | |
| N-[diethylamino(dimethyl)silyl]-N-ethylethanamine |
Specifications
| 4669-59-4 | |
| 193°C | |
| MFCD00039879 | |
| 1993 | |
| XIFOKLGEKUNZTI-UHFFFAOYSA-N | |
| N-[diethylamino(dimethyl)silyl]-N-ethylethanamine | |
| 78399 | |
| ≥98.0% (GC) | |
| Bis(diethylamino)dimethylsilane |
| Yellow | |
| C10H26N2Si | |
| 5 mL | |
| N,N,N′,N′-Tetraethyl-1,1-dimethylsilanediamine | |
| CCN(CC)[Si](C)(C)N(CC)CC | |
| 202.417 | |
| 202.42 | |
| Liquid |
Safety and Handling
TSCA : Yes