missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(diethylamino)dimethylsilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 193°C |
| Quantity | 5 mL |
| UN Number | 1993 |
| Formula Weight | 202.42 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(diethylamino)dimethylsilane |
Safety and Handling
| TSCA | Yes |